missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cycloheptene, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 304260050

 View more versions of this product

Catalog No. AC304260050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cycloheptene
628-92-2
Colorless
114.6°C
Authentic
Glass bottle
1.4570 to 1.459
MFCD00004156
05, 65
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
ZXIJMRYMVAMXQP-UHFFFAOYSA-N
cycloheptene
12363
95%
90%
-56°C
0.8200g/mL
-006°C
94% min. (GC)
C7H12
C7H12
5 g
0.82
Solubility in water: immiscible with water. Other solubilities: soluble in ether,ethanol,benzene
C1CCC=CCC1
96.17
96.17
Liquid

Chemical Identifiers

628-92-2
96.17
ZXIJMRYMVAMXQP-UHFFFAOYSA-N
12363
C1CCC=CCC1
C7H12
MFCD00004156
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
cycloheptene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-060-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only