Learn More
Cyclohexane (Certified ACS), Fisher Chemical™
$318.73 - $730.73
Chemical Identifiers
| CAS | 110-82-7 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00003814 |
| InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
| Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
| PubChem CID | 8078 |
| ChEBI | CHEBI:29005 |
| IUPAC Name | cyclohexane |
| SMILES | C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
C556-1
|
Fisher Chemical
BPC5561 |
1 L | Glass Bottle |
Each for $318.73
Case of 6 Each for $1,912.38
|
|
||||
|
C556-4
|
Fisher Chemical
BPC5564 |
4 L | Glass Bottle |
Each for $730.73
Case of 4 Each for $2,922.92
|
|
||||
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Specifications
| 6.5°C | |
| Colorless | |
| 81°C | |
| Pass Test | |
| Liquid | |
| ≥99.0 % | |
| CH2(CH2)4CH2 | |
| 104mbar at 20°C | |
| 0.02% max. | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.16 | |
| Certified ACS Reagent |
| 110-82-7 | |
| 0.770g/cm³ | |
| 1 L | |
| Glass Bottle | |
| 0.002% max. | |
| C6H12 | |
| MFCD00003814 | |
| 0.94 mPa/s at 20°C | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99.0% | |
| Cyclohexane |
Safety and Handling
DANGER!
Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid