missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥99.7%
$277.00 - $2215.88
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
6000471
|
Honeywell-Chromasolv
348551L |
1 L | Glass Bottle |
Each for $277.00
|
|
|||||
6000472
|
Honeywell-Chromasolv
3485525L |
2.5 L | Glass Bottle |
Each for $607.83
|
|
|||||
6000475
|
Honeywell-Chromasolv
348554X25L |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $2,097.23
|
|
|||||
6000476
|
Honeywell Chemicals
348554X4L |
4 x 4 L | Glass Bottle |
Case of 4 Each for $2,215.88
|
|
|||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Specifications
4°C to 7°C (literature) | |
max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm | |
0.779Â g/mLÂ at 25Â °C(lit.); 0.780Â g/mLÂ at 25Â °C | |
1 L | |
Glass Bottle | |
C6H12 | |
UN1145 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
HPLC |
110-82-7 | |
≤100.0000% | |
81°C | |
−18°C | |
C6H12 | |
MFCD00003814 | |
1900225 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
≥99.7% | |
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7% |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane
EINECSNumber : 203-806-2
Recommended Storage : Room Temp