missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, Environmental Grade, 99.7+%, Thermo Scientific Chemicals

Suitable for pesticide residue, environmental and GC analysis

Supplier:  Thermo Scientific Chemicals 040975K7

 View more versions of this product

Catalog No. AA40975K7


Explore available promotions

Description

Description

Used in environmental and GC analysis, for industrial production of adipic acid and caprolactam, which is a precursor of nylon, used as a solvent, and to make nylon and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexane
110-82-7
0.779 g/mL
4 L
Pungent
1.426
MFCD00003814
1900225
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
Environmental
6.5°C
Colorless
80°C to 81°C
-18°C (-1°F)
Liquid
C6H12
UN1145
14,2723
Immiscible in water and soluble in ether,alcohol and acetone. Miscible with olive oil.
C1CCCCC1
84.16
CHEBI:29005
≥99.7%

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H320-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXANE

EINECSNumber : 203-806-2

RTECSNumber : GU6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only