missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, for HPLC, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 610040040

Catalog No. AC610040040



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
Specifications

Specifications

Cyclohexane
110-82-7
2.90 (Air = 1.0)
Pass Test
10ppm max.
C6H12
Vapor Pressure: 104mbar at 20°C
05, 20
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.16
CHEBI:29005
≥99.0%
99.0%
(1 cm cell vs water), 0.014 max. at 254nm, 0.67 max. at 210nm, 0.88 max. at 205nm, 1 max. at 202nm
4 L
1.4235 - 1.4255
C6H12
MFCD00003814
0.02% max.
15, 2716
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.15
HPLC

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only