missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexane, Reagent, ACS, 99%, Spectrum™ Chemical
C1455, 110-82-7, C6H12
$204.38 - $2222.09
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18601943
|
Spectrum Chemical Mfg Cor
C1455100ML |
100 mL | Amber Glass Bottle |
Each for $204.38
|
|
||||
18601944
|
Spectrum Chemical Mfg Cor
C1455500MLGL |
500 mL | Amber Glass Bottle |
Each for $365.24
|
|
||||
18601946
|
Spectrum Chemical Mfg Cor
C14551LT |
1 L | Amber Glass Bottle |
Each for $461.50
|
|
||||
18601945
|
Spectrum Chemical Mfg Cor
C1455500MLGLCS6 |
6 x 500 mL | Amber Glass Bottle |
Case of 6 Each for $1,470.32
|
|
||||
18601947
|
Spectrum Chemical Mfg Cor
C14554LTGL |
4 L | Amber Glass Bottle |
Each for $763.00
|
|
||||
18601948
|
Spectrum Chemical Mfg Cor
C14551LTCS6 |
6 x 1 L | Amber Glass Bottle |
Case of 6 Each for $1,577.09
|
|
||||
18601949
|
Spectrum Chemical Mfg Cor
C14554LTGLCS4 |
4 x 4 L | Amber Glass Bottle |
Case of 4 Each for $2,222.09
|
|
||||
18601950
|
Spectrum Chemical Mfg Cor
C145520LTMT |
20 L | Steel Can |
Each for $2,192.00
|
|
||||
Description
Spectrum™ Chemical Cyclohexane, Reagent, ACS is used as a nonpolar solvent, in the calibration of differential scanning calorimetry and other industrial applications including a chemical intermediate. The distinctive detergent-like odor of cyclohexane is reminiscent of cleaning product and is sometimes used in cleaning products as a fragrance enhancer.Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
C1CCCCC1 |
C6H12 | |
MFCD00003814 | |
cyclohexane |
Specifications
110-82-7 | |
100 mL | |
0.002% | |
MFCD00003814 | |
0.02% | |
C1CCCCC1 | |
84.16 | |
ACS |
100% | |
Amber Glass Bottle | |
C6H12 | |
UN1145 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
99% |