missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexane (Spectranalyzed™), Fisher Chemical™
For use in the ultraviolet and infrared ranges
Supplier: Thermo Fisher Scientific FLC5554
Specifications
Cyclohexane | |
110-82-7 | |
Colorless | |
81°C | |
Pass Test | |
Amber Glass | |
Also meets ACS specifications | |
≥99.0 % | |
CH2(CH2)4CH2 | |
104mbar at 20°C | |
1145 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
≥99.0% |
6.5°C | |
Pass Test | |
0.770g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
0.002% max. | |
C6H12 | |
MFCD00003814 | |
0.94 mPa/s at 20°C | |
0.02% max. | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.16 | |
Spectranalyzed™ |
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid