Learn More
Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals
$51.14 - $147.01
Chemical Identifiers
CAS | 2719-27-9 |
---|---|
Molecular Formula | C7H11ClO |
Molecular Weight (g/mol) | 146.61 |
MDL Number | MFCD00001456 |
InChI Key | RVOJTCZRIKWHDX-UHFFFAOYSA-N |
Synonym | hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride |
PubChem CID | 75938 |
IUPAC Name | cyclohexanecarbonyl chloride |
SMILES | ClC(=O)C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1982414
|
Thermo Scientific Chemicals
A1982414 |
25 g |
Each for $51.14
|
|
|||||
AAA1982422
|
Thermo Scientific Chemicals
A1982422 |
100 g |
Each for $147.01
|
|
|||||
Description
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
2719-27-9 | |
146.61 | |
RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
75938 | |
ClC(=O)C1CCCCC1 |
C7H11ClO | |
MFCD00001456 | |
hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
cyclohexanecarbonyl chloride |
Specifications
2719-27-9 | |
183°C to 185°C | |
C7H11ClO | |
MFCD00001456 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
ClC(=O)C1CCCCC1 | |
146.61 | |
146.62 | |
Cyclohexanecarbonyl chloride |
1.088 | |
66°C (150°F) | |
1.47 | |
25 g | |
742163 | |
hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
cyclohexanecarbonyl chloride | |
75938 | |
≥97% |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-322-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only