missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

$75.66 - $129.23

Chemical Identifiers

CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1469330
View Documents
Thermo Scientific Chemicals
A1469330
250 g
Each for $75.66
 
AAA1469336
View Documents
Thermo Scientific Chemicals
A1469336
500 g
Each for $129.23
 
Description

Description

Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

Solubility
Soluble in water 0.201 g/100 mL (15°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

98-89-5
128.171
NZNMSOFKMUBTKW-UHFFFAOYSA-N
7413
cyclohexanecarboxylic acid
C7H12O2
MFCD00001461
hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
CHEBI:36096
C1CCC(CC1)C(=O)O

Specifications

98-89-5
1.048
118°C (244°F)
1.461
250 g
14,2724
Soluble in water 0.201g/100 mL (15°C).
C1CCC(CC1)C(=O)O
128.171
CHEBI:36096
98%
29°C to 32°C
231°C to 233°C
C7H12O2
MFCD00001461
970529
hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
NZNMSOFKMUBTKW-UHFFFAOYSA-N
cyclohexanecarboxylic acid
7413
128.17
Cyclohexanecarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-711-3

RTECSNumber : GU8370000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only