missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C146220LTMT

 View more versions of this product

Catalog No. 18610102


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cyclohexanone
1
20 L
0.0005
0.99
108-94-1
156°C
Steel Can
0.0005
Reagent

Chemical Identifiers

108-94-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.