missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C1462200LTBL

 View more versions of this product

Catalog No. 18610103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Cyclohexanone
1
200 L
0.0005
0.99
108-94-1
156°C
Drum
0.0005
Reagent

Chemical Identifiers

108-94-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.