missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanone cyanohydrin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2669909

 View more versions of this product

Catalog No. AAH2669909


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexanone cyanohydrin
34°C to 36°C
132°C (19 mmHg)
C7H11NO
MFCD00003818
UN3439
ZDBRPNZOTCHLSP-UHFFFAOYSA-N
1-hydroxycyclohexane-1-carbonitrile
13610
98%
931-97-5
1.031
118°C (244°F)
1.469
10 g
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
C1CCC(CC1)(C#N)O
125.171
125.17

Chemical Identifiers

931-97-5
125.171
ZDBRPNZOTCHLSP-UHFFFAOYSA-N
13610
C1CCC(CC1)(C#N)O
C7H11NO
MFCD00003818
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
1-hydroxycyclohexane-1-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P501c

H300+H310+H330-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 213-246-0

RTECSNumber : GU7710000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only