Learn More
Cyclohexanone cyanohydrin, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2669918
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclohexanone cyanohydrin | |
34°C to 36°C | |
132°C (19 mmHg) | |
C7H11NO | |
MFCD00003818 | |
UN3439 | |
ZDBRPNZOTCHLSP-UHFFFAOYSA-N | |
1-hydroxycyclohexane-1-carbonitrile | |
13610 | |
98% |
931-97-5 | |
1.031 | |
118°C (244°F) | |
1.469 | |
50 g | |
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin | |
C1CCC(CC1)(C#N)O | |
125.171 | |
125.17 |
Chemical Identifiers
931-97-5 | |
125.171 | |
ZDBRPNZOTCHLSP-UHFFFAOYSA-N | |
13610 | |
C1CCC(CC1)(C#N)O |
C7H11NO | |
MFCD00003818 | |
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin | |
1-hydroxycyclohexane-1-carbonitrile |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P501c
H300+H310+H330-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 213-246-0
RTECSNumber : GU7710000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only