missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanone Oxime 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111201000

 View more versions of this product

Catalog No. AC111201000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexanone oxime
97%
Beige-Brown to White
Authentic
Crystalline Powder, Crystals, or Chunks
C6H11NO
MFCD00001660
10, 2720
VEZUQRBDRNJBJY-UHFFFAOYSA-N
N-cyclohexylidenehydroxylamine
7517
≥96.0%
88°C to 92°C
100-64-1
100 g
Plastic Bottle
97%
C6H10(=NOH)
07, 10
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
ON=C1CCCCC1
113.16
113.16

Chemical Identifiers

100-64-1
113.16
VEZUQRBDRNJBJY-UHFFFAOYSA-N
7517
ON=C1CCCCC1
C6H11NO
MFCD00001660
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
N-cyclohexylidenehydroxylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-874-0

RTECSNumber : GW1925000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only