missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Cycloheximide, 95%

Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.

$144.94 - $1511.58

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC357420010
SDS View Documents
Thermo Scientific Chemicals
357420010
1 g
Each for $144.94
 
AC357420050
SDS View Documents
Thermo Scientific Chemicals
357420050
5 g
Each for $357.32
 
AC357420250
SDS View Documents
Thermo Scientific Chemicals
357420250
25 g
Each for $1,511.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cycloheximide is a semisynthetic compound showing an antifungal antibiotic activity.
  • It is produced by the bacterium Streptomyces griseus
  • Cycloheximide can inhibit protein synthesis in eukaryotic cells but not in prokaryotes by interacting directly with the translocase enzyme and interfering with the translocation step. Furthermore, this compound also blocks the A site of ribosomes

Application

  • Cycloheximide shows a neuroprotective, fungicide, and anticoronaviral activities
  • In biomedical research, it can induce glycogenolysis, gluconeogenesis, and ureagenesis
  • Due to significant toxic side effects such as teratogenesis, DNA damage, birth defects, and toxicity to sperm, this chemical compound is only used in in vitro research applications
  • This compound also shows inhibitory activity against ferroptosis
  • It is also used in laboratory media as a selective agent to permit the isolation of pathogenic and non-pathogenic fungi from other types of specimens
  • In laboratory research related to plant biology, it can induce ethylene production to promote the plant growth
  • In laboratory research, this compound can be applied to determine the half-life of protein in any cellular system. The treatment of in vitro cells with cycloheximide in a time-course experiment followed by western blotting using this cell lysate can show differences in protein half-life.
Specifications

Chemical Identifiers

66-81-9
281.35
YPHMISFOHDHNIV-QTEFRXOUNA-N
6197
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
C15H23NO4
MFCD00082346
cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
CHEBI:27641
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

66-81-9
White
94% min. (HPLC)
C15H23NO4
1 g
− 28.50 (20.00°C c=1,CHCl3)
cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
YPHMISFOHDHNIV-QTEFRXOUNA-N
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
6197
281.35
Glassy Irregular Shaped Granules
107.0°C to 114.0°C
Authentic
Glass bottle
MFCD00082346
15,2721
− 28.50
Solubility in water: 2.1 g/100mL (2 c). Other solubilities: soluble in chloroform, ether, acetone, methanol, and ethanol
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
281.35
CHEBI:27641
95%
Cycloheximide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Suspected of causing genetic defects.
May damage the unborn child.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-636-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only