Learn More
Cyclohexyl Bromide, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 159080010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclohexyl bromide | |
1.3300g/mL | |
62°C | |
99% | |
C6H11Br | |
C6H11Br | |
1L | |
1.33 | |
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
bromocyclohexane | |
7960 | |
99% |
108-85-0 | |
166.0°C to 167.0°C | |
Authentic | |
Glass bottle | |
1.4940 to 1.4960 | |
MFCD00003819 | |
05, 24 | |
15, 2723 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
BrC1CCCCC1 | |
163.06 | |
163.06 |
Chemical Identifiers
108-85-0 | |
163.06 | |
AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
7960 | |
BrC1CCCCC1 |
C6H11Br | |
MFCD00003819 | |
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
bromocyclohexane |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
In use, may form flammable/explosive vapor-air mixture.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 203-622-2
RUO â Research Use Only