missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111300010

 View more versions of this product

Catalog No. AC111300010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexyl chloride
542-18-7
1.0000g/mL
28°C
97.5% min. (GC)
C6H11Cl
C6H11Cl
1 kg
1
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
UNFUYWDGSFDHCW-UHFFFAOYSA-N
chlorocyclohexane
10952
CHEBI:39156
98%
98%
-44°C
142°C
Authentic
Glass bottle
1.4610 to 1.463
MFCD00003822
05, 21
15, 2725
Solubility in water: 0.02g/L (20°C). Other solubilities: soluble in alcohol and diethyl ether
C1CCC(CC1)Cl
118.61
1.73 mPa.s (20°C)
118.61
Liquid

Chemical Identifiers

542-18-7
118.61
UNFUYWDGSFDHCW-UHFFFAOYSA-N
10952
chlorocyclohexane
C6H11Cl
MFCD00003822
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
CHEBI:39156
C1CCC(CC1)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-806-6

RTECSNumber : GU8685000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only