Learn More
Cyclohexyl isocyanide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158400050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclohexyl isocyanide | |
Brown to Colorless | |
173.0°C to 176.0°C | |
Authentic | |
Glass bottle | |
1.4490 to 1.4520 | |
MFCD00003839 | |
06,152 | |
cyclohexyl isocyanide, cyclohexane, isocyano, cyclohexylisocyanide, cyclohexaneisonitrile, cyclohexyl isonitrile, n-cyclohexylisocyanide, cyi, cyclohexylisonitrile, isocyano-cyclohexane, cyclohexyliminocarbene | |
XYZMOVWWVXBHDP-UHFFFAOYSA-N | |
isocyanocyclohexane | |
79129 | |
109.17 | |
Liquid |
931-53-3 | |
0.8900g/mL | |
77°C | |
98.5% min. (GC) | |
C7H11N | |
C6H11NC | |
5 g | |
0.89 | |
Solubility in water: insoluble. Other solubilities: soluble in inert organic solvents | |
[C-]#[N+]C1CCCCC1 | |
109.17 | |
CHEBI:17966 | |
99% |
Chemical Identifiers
931-53-3 | |
109.17 | |
XYZMOVWWVXBHDP-UHFFFAOYSA-N | |
79129 | |
isocyanocyclohexane |
C7H11N | |
MFCD00003839 | |
cyclohexyl isocyanide, cyclohexane, isocyano, cyclohexylisocyanide, cyclohexaneisonitrile, cyclohexyl isonitrile, n-cyclohexylisocyanide, cyi, cyclohexylisonitrile, isocyano-cyclohexane, cyclohexyliminocarbene | |
CHEBI:17966 | |
[C-]#[N+]C1CCCCC1 |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 213-238-7
RUO – Research Use Only