missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine Hydrochloride, 99%, Spectrum™ Chemical

$169.85 - $11703.76

Chemical Identifiers

CAS 4998-76-9
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610120
View Documents
Spectrum Chemical Mfg Cor
C212725GM
25 g Amber Glass Bottle
Each for $169.85
Only null left
 
18610117
View Documents
Spectrum Chemical Mfg Cor
C2127100GM
100 g Amber Glass Bottle
Each for $336.68
Only null left
 
18610121
View Documents
Spectrum Chemical Mfg Cor
C2127500GM
500 g Amber Glass Bottle
Each for $606.12
Only null left
 
18610119
View Documents
Spectrum Chemical Mfg Cor
C212725KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4998-76-9

Specifications

4998-76-9
25 g
Reagent
1
99%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.