missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexylamine hydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 134655000

 View more versions of this product

Catalog No. AC134655000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclohexylamine hydrochloride
4998-76-9
Authentic
Plastic bottle
MFCD00050540
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexanamine;hydrochloride
2723770
99%
99%
306.0°C
99%
C6H14ClN
500g
Solubility in water: 83 g/100mL water (17°C). Other solubilities: soluble in most common organic solvents, practically insoluble in ethers
[Cl-].[NH3+]C1CCCCC1
135.64
135.63

Chemical Identifiers

4998-76-9
135.64
ZJUGSKJHHWASAF-UHFFFAOYSA-N
2723770
[Cl-].[NH3+]C1CCCCC1
C6H14ClN
MFCD00050540
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
cyclohexanamine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Suspected of damaging fertility.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 225-661-4

RTECSNumber : GX1440000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only