missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmagnesium chloride, 1M in MeTHF

CAS: 931-51-1 | C6H11ClMg | 142.91 g/mol

$103.11 - $406.01

Chemical Identifiers

CAS 931-51-1
Molecular Formula C6H11ClMg
Molecular Weight (g/mol) 142.91
MDL Number MFCD00003816
InChI Key WMJMABVHDMRMJA-UHFFFAOYSA-M
Synonym cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
PubChem CID 13605
IUPAC Name magnesium;cyclohexane;chloride
SMILES Cl[Mg]C1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51165AE
View Documents
Thermo Scientific Chemicals
H51165AE
100 mL N/A
Only null left
 
AAH51165AP
View Documents
Thermo Scientific Chemicals
H51165AP
500 mL N/A
Only null left
 
Description

Description

Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

931-51-1
142.91
WMJMABVHDMRMJA-UHFFFAOYSA-M
13605
Cl[Mg]C1CCCCC1
C6H11ClMg
MFCD00003816
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
magnesium;cyclohexane;chloride

Specifications

931-51-1
MFCD00003816
UN2924
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
WMJMABVHDMRMJA-UHFFFAOYSA-M
magnesium;cyclohexane;chloride
13605
1M in MeTHF
C6H11ClMg
100 mL
Air and moisture sensitive
It reacts violently with water.
Cl[Mg]C1CCCCC1
142.91
142.91
Cyclohexylmagnesium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only