Learn More
(Cyclohexylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H58186AD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(Cyclohexylmethyl)zinc bromide | |
135579-86-1 | |
C7H13BrZn | |
50 mL | |
Air sensitive | |
SJCALMUTSZRVSS-UHFFFAOYSA-M | |
bromozinc(1+);methanidylcyclohexane | |
4486981 |
Packaged under Argon in resealable ChemSeal™ bottles | |
0.972 | |
MFCD01311418 | |
UN3399 | |
cyclohexylmethyl zinc bromide solution, cyclohexyl methylzinc bromide, cyclohexylmethyl zinc ii bromide, bromozinc 1+ ,methanidylcyclohexane, cyclohexylmethyl zinc bromide, 0.50 m in thf, cyclohexyl methylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylmethyl zinc bromide solution, 0.5 m in thf, cyclohexylmethyl zinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles | |
[CH2-]C1CCCCC1.[Zn+]Br | |
242.465 | |
242.46 |
Chemical Identifiers
135579-86-1 | |
242.465 | |
SJCALMUTSZRVSS-UHFFFAOYSA-M | |
4486981 | |
[CH2-]C1CCCCC1.[Zn+]Br |
C7H13BrZn | |
MFCD01311418 | |
cyclohexylmethyl zinc bromide solution, cyclohexyl methylzinc bromide, cyclohexylmethyl zinc ii bromide, bromozinc 1+ ,methanidylcyclohexane, cyclohexylmethyl zinc bromide, 0.50 m in thf, cyclohexyl methylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylmethyl zinc bromide solution, 0.5 m in thf, cyclohexylmethyl zinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles | |
bromozinc(1+);methanidylcyclohexane |
Safety and Handling
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only