missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclooctane, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L15000AD

 View more versions of this product

Catalog No. AAL15000AD


Explore available promotions

Description

Description

Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications

Specifications

Cyclooctane
12°C to 14°C
150°C to 152°C
C8H16
MFCD00004162
UN3295
octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
WJTCGQSWYFHTAC-UHFFFAOYSA-N
cyclooctane
9266
≥99%
292-64-8
0.835
28°C (82°F)
1.458
50 mL
1900349
Not miscible or difficult to mix in water.
C1CCCCCCC1
112.216
112.22

Chemical Identifiers

292-64-8
112.216
WJTCGQSWYFHTAC-UHFFFAOYSA-N
9266
C1CCCCCCC1
C8H16
MFCD00004162
octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
cyclooctane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 206-031-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only