Learn More
Cyclooctanol, 97%, Thermo Scientific Chemicals
$138.79 - $412.71
Chemical Identifiers
CAS | 696-71-9 |
---|---|
Molecular Formula | C8H16O |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00001744 |
InChI Key | FHADSMKORVFYOS-UHFFFAOYSA-N |
Synonym | cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa |
PubChem CID | 12766 |
IUPAC Name | cyclooctanol |
SMILES | OC1CCCCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1782514
|
Thermo Scientific Chemicals
A1782514 |
25 g |
Each for $138.79
|
|
||||||
AAA1782522
|
Thermo Scientific Chemicals
A1782522 |
100 g |
Each for $412.71
|
|
||||||
Description
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
Solubility
Miscible with acetone.
Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
Chemical Identifiers
696-71-9 | |
128.22 | |
FHADSMKORVFYOS-UHFFFAOYSA-N | |
12766 | |
OC1CCCCCCC1 |
C8H16O | |
MFCD00001744 | |
cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa | |
cyclooctanol |
Specifications
696-71-9 | |
0.97 | |
86°C (186°F) | |
1.486 | |
25 g | |
cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa | |
FHADSMKORVFYOS-UHFFFAOYSA-N | |
cyclooctanol | |
12766 | |
97% |
14°C to 15°C | |
97°C to 98°C (11 mmHg) | |
C8H16O | |
MFCD00001744 | |
1848166 | |
Miscible with acetone. | |
OC1CCCCCCC1 | |
128.22 | |
128.22 | |
Cyclooctanol |
Safety and Handling
GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 211-800-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only