Learn More
Cyclooctanone, 98%, Thermo Scientific Chemicals
$68.95 - $628.99
Chemical Identifiers
CAS | 502-49-8 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00001754 |
InChI Key | IIRFCWANHMSDCG-UHFFFAOYSA-N |
Synonym | cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference |
PubChem CID | 10403 |
IUPAC Name | cyclooctanone |
SMILES | C1CCCC(=O)CCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2091114
|
Thermo Scientific Chemicals
B2091114 |
25 g |
Each for $68.95
|
|
|||||
AAB2091122
|
Thermo Scientific Chemicals
B2091122 |
100 g |
Each for $190.71
|
|
|||||
AAB2091136
|
Thermo Scientific Chemicals
B2091136 |
500 g |
Each for $628.99
|
|
|||||
Description
Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Chemical Identifiers
502-49-8 | |
126.199 | |
IIRFCWANHMSDCG-UHFFFAOYSA-N | |
10403 | |
C1CCCC(=O)CCC1 |
C8H14O | |
MFCD00001754 | |
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
cyclooctanone |
Specifications
502-49-8 | |
0.96 | |
72°C (161°F) | |
C8H14O | |
25 g | |
1280738 | |
Soluble in water at 20°C 15g/L. Soluble in acetone,alcohol,chloroform,methanol and benzene. | |
C1CCCC(=O)CCC1 | |
126.199 | |
126.2 | |
Cyclooctanone |
37°C to 44°C | |
195°C to 197°C | |
Pleasant | |
MFCD00001754 | |
UN1759 | |
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
IIRFCWANHMSDCG-UHFFFAOYSA-N | |
cyclooctanone | |
10403 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 207-940-2
RTECSNumber : GX9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only