missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopenta[fg]tetracene-1,2-dione, TCI America™

$304.49 - $1,058.83

Chemical Identifiers

CAS 1334510-66-5
Molecular Formula C20H10O2
Molecular Weight (g/mol) 282.298
InChI Key AYSSMZWWOZPRMT-UHFFFAOYSA-N
Synonym Cyclopenta[fg]naphthacene-1,2-dione
PubChem CID 91972158
SMILES C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C2669200MG
View Documents
TCI America
C2669200MG
200 mg N/A
 
C26691G
View Documents
TCI America
C26691G
1 g N/A
 
Description

Description

Specifications

Chemical Identifiers

1334510-66-5
282.298
Cyclopenta[fg]naphthacene-1,2-dione
C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O
C20H10O2
AYSSMZWWOZPRMT-UHFFFAOYSA-N
91972158

Specifications

1334510-66-5
C20H10O2
Cyclopenta[fg]naphthacene-1,2-dione
C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O
91972158
Crystalline Powder
Blue-Purple
200 mg
AYSSMZWWOZPRMT-UHFFFAOYSA-N
282.298
282.30
Cyclopenta[fg]tetracene-1,2-dione
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'