missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentane, 95+%, pure, Thermo Scientific Chemicals

Cyclopentane, 95+%, C5H10, CAS Number-287-92-3, pentamethylene, cyclopentanes, hsdb 62, hydrocarbons, cyclic c5 and c6, cyclopentan, t86pb90rnu, unii-t86pb90rnu, ciclopentano, cyclopentyl group, zyklopentan, 5L, 95% min. (GC), 50.0 deg.C, CHEBI:23492, Colorless, 0.7500g/mL, 206-016-6, -7 deg.C

Supplier:  Thermo Scientific Chemicals 397430010

 View more versions of this product

Catalog No. AC397430010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentane
287-92-3
Colorless
50.0°C
Authentic
Glass Bottle
1.4050 to 1.4070
MFCD00001356
0.001% max.
15, 2736
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents
C1CCCC1
70.15
0.44 mPa.s (20°C)
70.15
Pure
Extra Dry
-94.0°C
0.7500g/mL
−7°C
95% min. (GC)
C5H10
C5H10
1 L
0.75
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
RGSFGYAAUTVSQA-UHFFFAOYSA-N
cyclopentane
9253
CHEBI:23492
95+%
Liquid

Chemical Identifiers

287-92-3
70.15
RGSFGYAAUTVSQA-UHFFFAOYSA-N
9253
cyclopentane
C5H10
MFCD00001356
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
CHEBI:23492
C1CCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 206-016-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only