missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentanecarbonyl chloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 354371000

 View more versions of this product

Catalog No. AC354371000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclopentanecarbonyl chloride
1.0910g/mL
60°C
98%
C6H9ClO
100g
cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
C1CCC(C1)C(=O)Cl
132.59
132.59
4524-93-0
54.0°C to 56.0°C (15.0mmHg)
Authentic
Glass bottle
1.4610 to 1.4640
1.091
WEPUZBYKXNKSDH-UHFFFAOYSA-N
cyclopentanecarbonyl chloride
78284
98%

Chemical Identifiers

4524-93-0
132.59
cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
cyclopentanecarbonyl chloride
C6H9ClO
WEPUZBYKXNKSDH-UHFFFAOYSA-N
78284
C1CCC(C1)C(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 224-856-1

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only