Learn More
Cyclopentanecarboxaldehyde, 97%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 425910050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopentanecarboxaldehyde | |
0.9190g/mL | |
28°C | |
96% min. (GC) | |
C6H10O | |
C5H9CHO | |
0.919 | |
VELDYOPRLMJFIK-UHFFFAOYSA-N | |
cyclopentanecarbaldehyde | |
0.1 wt% hydroquinone | |
98.14 |
872-53-7 | |
140.0°C to 141.0°C | |
Authentic | |
Glass bottle | |
1.4420 to 1.4460 (20°C, 589nm) | |
5g | |
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
C1CCC(C1)C=O | |
98.14 | |
70106 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 212-829-7
RUO â Research Use Only