missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde, 97%, stabilized

CAS: 872-53-7 | C6H10O | 98.14 g/mol

$852.35 - $852.35

Chemical Identifiers

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC425910250
SDS View Documents
Thermo Scientific Chemicals
425910250
25 g Glass bottle
Each for $852.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

872-53-7
98.14
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
cyclopentanecarbaldehyde
C6H10O
VELDYOPRLMJFIK-UHFFFAOYSA-N
70106
C1CCC(C1)C=O

Specifications

872-53-7
0.9190g/mL
28°C
96% min. (GC)
C6H10O
C5H9CHO
0.919
VELDYOPRLMJFIK-UHFFFAOYSA-N
cyclopentanecarbaldehyde
0.1 wt% hydroquinone
98.14
Liquid
Colorless to Yellow
140.0°C to 141.0°C
Authentic
Glass bottle
1.4420 to 1.4460 (20°C, 589nm)
25 g
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
C1CCC(C1)C=O
98.14
70106
97%
Cyclopentanecarboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 212-829-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only