missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111490250

 View more versions of this product

Catalog No. AC111490250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentanecarboxylic acid
3400-45-1
Colorless to Yellow
216.0°C
Authentic
Glass bottle
1.4530 to 1.5450
MFCD00001371
09, 6
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
JBDSSBMEKXHSJF-UHFFFAOYSA-N
cyclopentanecarboxylic acid
18840
98+%
98+%
4.0°C
1.0500g/mL
93°C
98% min. (GC)
C6H10O2
C5H9CO2H
25 g
1.05
Solubility in water: 3g/100mL. Other solubilities: soluble in alcohols, methylene chloride, acetone,, toluene
C1CCC(C1)C(=O)O
114.14
114.14
Liquid

Chemical Identifiers

3400-45-1
114.14
JBDSSBMEKXHSJF-UHFFFAOYSA-N
18840
C1CCC(C1)C(=O)O
C6H10O2
MFCD00001371
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
cyclopentanecarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-269-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only