missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A152490F

 View more versions of this product

Catalog No. AAA152490F


Explore available promotions

Description

Description

Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Cyclopentanol
-19°C
139°C to 141°C
Unpleasant
1.453
2500 mL
1900556
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
XCIXKGXIYUWCLL-UHFFFAOYSA-N
cyclopentanol
7298
86.13
96-41-3
0.95
51°C (123°F)
C5H10O
MFCD00001363
UN2244
14,2742
Slightly soluble in water. Also soluble in ether,alcohols and acetone.
C1CCC(C1)O
86.134
CHEBI:16133
99%

Chemical Identifiers

96-41-3
86.134
XCIXKGXIYUWCLL-UHFFFAOYSA-N
7298
cyclopentanol
C5H10O
MFCD00001363
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
CHEBI:16133
C1CCC(C1)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P243-P261-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P374-P380-P501c

H226-H302-H310-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANOL

EINECSNumber : 202-504-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only