Learn More
Cyclopentanol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A15249AP
Description
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopentanol is a versatile compound used as a building block, intermediate and solvent.
Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Cyclopentanol | |
-19°C | |
139°C to 141°C | |
Unpleasant | |
1.453 | |
500 mL | |
1900556 | |
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol | |
XCIXKGXIYUWCLL-UHFFFAOYSA-N | |
cyclopentanol | |
7298 | |
86.13 |
96-41-3 | |
0.95 | |
51°C (123°F) | |
C5H10O | |
MFCD00001363 | |
UN2244 | |
14,2742 | |
Slightly soluble in water. Also soluble in ether,alcohols and acetone. | |
C1CCC(C1)O | |
86.134 | |
CHEBI:16133 | |
99% |
Chemical Identifiers
96-41-3 | |
86.134 | |
XCIXKGXIYUWCLL-UHFFFAOYSA-N | |
7298 | |
cyclopentanol |
C5H10O | |
MFCD00001363 | |
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol | |
CHEBI:16133 | |
C1CCC(C1)O |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P243-P261-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P374-P380-P501c
H226-H302-H310-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANOL
EINECSNumber : 202-504-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only