missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentanone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A14222AK

 View more versions of this product

Catalog No. AAA14222AK


Explore available promotions

Description

Description

Cyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong reducing agents, strong oxidizing agents and strong bases.
Specifications

Specifications

Cyclopentanone
-51°C
130°C to 131°C
Petrol-like
1.437
250 mL
605573
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
BGTOWKSIORTVQH-UHFFFAOYSA-N
cyclopentanone
8452
84.12
120-92-3
0.949
30°C (86°F)
C5H8O
MFCD00001409
UN2245
14,2743
Slightly miscible with water.
O=C1CCCC1
84.12
CHEBI:16486
99%

Chemical Identifiers

120-92-3
84.12
BGTOWKSIORTVQH-UHFFFAOYSA-N
8452
cyclopentanone
C5H8O
MFCD00001409
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
CHEBI:16486
O=C1CCCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANONE

EINECSNumber : 204-435-9

RTECSNumber : GY4725000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only