missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopentanone oxime, 97%, Thermo Scientific Chemicals
$139.08 - $293.29
Chemical Identifiers
CAS | 1192-28-5 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.13 |
MDL Number | MFCD00001420 |
InChI Key | YGNXYFLJZILPEK-UHFFFAOYSA-N |
Synonym | cyclopentanone oxime, cyclopentanoneoxime, cyclopentanone, oxime, unii-qy3bv00pcy, ccris 7163, qy3bv00pcy, n-1e-cyclopentylidene hydroxylamine, hydroxyimino cyclopentane, cyclopentanone,oxime, hydroxyiminocyclopentane |
PubChem CID | 14500 |
IUPAC Name | N-cyclopentylidenehydroxylamine |
SMILES | ON=C1CCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2496114
|
Thermo Scientific Chemicals
B2496114 |
25 g |
Each for $139.08
|
|
|||||
AAB2496122
|
Thermo Scientific Chemicals
B2496122 |
100 g |
Each for $293.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1192-28-5 | |
99.13 | |
YGNXYFLJZILPEK-UHFFFAOYSA-N | |
14500 | |
ON=C1CCCC1 |
C5H9NO | |
MFCD00001420 | |
cyclopentanone oxime, cyclopentanoneoxime, cyclopentanone, oxime, unii-qy3bv00pcy, ccris 7163, qy3bv00pcy, n-1e-cyclopentylidene hydroxylamine, hydroxyimino cyclopentane, cyclopentanone,oxime, hydroxyiminocyclopentane | |
N-cyclopentylidenehydroxylamine |
Specifications
1192-28-5 | |
196°C | |
C5H9NO | |
25 g | |
YGNXYFLJZILPEK-UHFFFAOYSA-N | |
N-cyclopentylidenehydroxylamine | |
14500 | |
97% |
54°C to 58°C | |
92°C (198°F) | |
MFCD00001420 | |
cyclopentanone oxime, cyclopentanoneoxime, cyclopentanone, oxime, unii-qy3bv00pcy, ccris 7163, qy3bv00pcy, n-1e-cyclopentylidene hydroxylamine, hydroxyimino cyclopentane, cyclopentanone,oxime, hydroxyiminocyclopentane | |
ON=C1CCCC1 | |
99.13 | |
99.13 | |
Cyclopentanone oxime |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 214-749-8
RTECSNumber : GY5150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only