missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041461AE

 View more versions of this product

Catalog No. AA41461AE


Explore available promotions

Description

Description

Cyclopentene is used in organic synthesis for cross-linking resin. It is used as an intermediate in Industries such as Agrochemical, Dyestuff, Pharmaceutical, syntheses material, Chemical. It is used as a monomer for synthesis of plastics and rubber. It is also used for the synthesis of various chemicals, such as 1,1,2-Trimethylcyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentene
-135°C
0.773 g/mL
-29°C (-20°F)
1.4225
100 mL
635707
1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
LPIQUOYDBNQMRZ-UHFFFAOYSA-N
cyclopentene
8882
68.12
Liquid
142-29-0
Colorless
44°C to 45°C
C5H8
MFCD00001394
UN2246
Air Sensitive
It is soluble in most organic solvents like ethanol and ether. It is immiscible with water.
C1CC=CC1
68.119
CHEBI:49155
≥98%

Chemical Identifiers

142-29-0
68.119
LPIQUOYDBNQMRZ-UHFFFAOYSA-N
8882
cyclopentene
C5H8
MFCD00001394
1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
CHEBI:49155
C1CC=CC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H302-H312-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Harmful in contact with skin.
May cause drowsiness or dizziness.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P312-P330-P331-P332+P313-P363-P370+P378q-P501c

H225-H302+H312-H304-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOPENTENE

EINECSNumber : 205-532-9

RTECSNumber : GY5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only