missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopentene oxide, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 285-67-6 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.12 |
MDL Number | MFCD00005161 |
InChI Key | GJEZBVHHZQAEDB-UHFFFAOYNA-N |
Synonym | cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane |
PubChem CID | 9244 |
IUPAC Name | 6-oxabicyclo[3.1.0]hexane |
SMILES | C1CC2OC2C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC348520250
|
Thermo Scientific Chemicals
348520250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC348521000
|
Thermo Scientific Chemicals
348521000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
285-67-6 | |
84.12 | |
GJEZBVHHZQAEDB-UHFFFAOYNA-N | |
9244 | |
C1CC2OC2C1 |
C5H8O | |
MFCD00005161 | |
cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane | |
6-oxabicyclo[3.1.0]hexane |
Specifications
285-67-6 | |
102.0°C | |
Authentic | |
Glass bottle | |
1.4330 to 1.4350 | |
25g | |
0.964 | |
GJEZBVHHZQAEDB-UHFFFAOYNA-N | |
6-oxabicyclo[3.1.0]hexane | |
9244 | |
98% |
0.9640g/mL | |
10°C | |
97.5% min. (GC) | |
C5H8O | |
MFCD00005161 | |
17,21 | |
cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane | |
C1CC2OC2C1 | |
84.12 | |
84.12 | |
Cyclopentene oxide |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 206-005-6
RUO â Research Use Only