missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopentyl iodide, 98%, stabilized over copper, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 381730250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopentyl iodide | |
1556-18-9 | |
166.0°C to 167.0°C | |
Authentic | |
Glass bottle | |
C5H9I | |
25mL | |
cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu | |
IC1CCCC1 | |
196.03 | |
73784 | |
98% |
Stabilized | |
1.6950g/mL | |
52°C | |
96% min. (GC) | |
C5H9I | |
MFCD00039446 | |
1.695 | |
PCEBAZIVZVIQEO-UHFFFAOYSA-N | |
iodocyclopentane | |
(Copper) | |
196.03 |
Chemical Identifiers
1556-18-9 | |
196.03 | |
PCEBAZIVZVIQEO-UHFFFAOYSA-N | |
73784 | |
IC1CCCC1 |
C5H9I | |
MFCD00039446 | |
cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu | |
iodocyclopentane |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a
GHS Signal Word: Warning
EINECSNumber : 216-311-1
RUO â Research Use Only