missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isocyanide, 96%

CAS: 68498-54-4 | C6H9N | 95.14 g/mol

$336.73 - $336.73

Chemical Identifiers

CAS 68498-54-4
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.14
InChI Key XTLSDEJNOQVCHL-UHFFFAOYSA-N
Synonym cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
PubChem CID 2734784
IUPAC Name isocyanocyclopentane
SMILES [C-]#[N+]C1CCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC372090010
SDS View Documents
Thermo Scientific Chemicals
372090010
1 g Glass bottle
Each for $336.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68498-54-4
95.14
cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
isocyanocyclopentane
C6H9N
XTLSDEJNOQVCHL-UHFFFAOYSA-N
2734784
[C-]#[N+]C1CCCC1

Specifications

68498-54-4
0.8600g/mL
Authentic
Glass bottle
1.4390 to 1.4410
0.86
Solubility in water: insoluble
[C-]#[N+]C1CCCC1
95.14
95.14
Liquid
Colorless to Yellow
40°C (11.0 hPa)
95% min. (GC)
C6H9N
1 g
cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
XTLSDEJNOQVCHL-UHFFFAOYSA-N
isocyanocyclopentane
2734784
96%
Cyclopentyl isocyanide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only