missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentyl methyl ketone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6474406

 View more versions of this product

Catalog No. AAH6474406


Explore available promotions

Description

Description

Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1'-cyclopentan)-3-amin-1,4-dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1′-cyclopentan)-3-amin-1,4-dioxide.

Solubility
Miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, reducing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
Specifications

Specifications

Cyclopentyl methyl ketone
0.913
C7H12O
MFCD00060799
1853483
Miscible with water.
CC(=O)C1CCCC1
112.17
112.17
6004-60-0
151°C to 156°C
1.4435
5 g
1-cyclopentyl-ethanone, cyclopentyl methyl ketone, ethanone, 1-cyclopentyl, acetylcyclopentane, cyclopentylethanone, ketone, cyclopentyl methyl, 1-cyclopentylethan-1-one, ethanone, 1-cyclopentyl-9ci, ethanone,1-cyclopentyl, methyl cyclopentyl ketone
LKENTYLPIUIMFG-UHFFFAOYSA-N
1-cyclopentylethanone
22326
98%

Chemical Identifiers

6004-60-0
112.17
LKENTYLPIUIMFG-UHFFFAOYSA-N
22326
CC(=O)C1CCCC1
C7H12O
MFCD00060799
1-cyclopentyl-ethanone, cyclopentyl methyl ketone, ethanone, 1-cyclopentyl, acetylcyclopentane, cyclopentylethanone, ketone, cyclopentyl methyl, 1-cyclopentylethan-1-one, ethanone, 1-cyclopentyl-9ci, ethanone,1-cyclopentyl, methyl cyclopentyl ketone
1-cyclopentylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only