missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentylamine, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 111591000

Catalog No. AC111591000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclopentylamine
1003-03-8
106°C to 108°C
Authentic
Glass bottle
1.4482 to 1.4502
MFCD00001380
12,4
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
NISGSNTVMOOSJQ-UHFFFAOYSA-N
cyclopentanamine
2906
99+%
99+%
0.8600g/mL
17°C
99% min. (GC)
C5H11N
C5H9NH2
100g
0.86
Solubility in water: completely miscible with water
NC1CCCC1
85.15
85.15
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-697-3

RTECSNumber : GY8452000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only