missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentylamine, 99%, Thermo Scientific Chemicals

$76.63 - $204.41

Chemical Identifiers

CAS 1003-03-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00001380
InChI Key NISGSNTVMOOSJQ-UHFFFAOYSA-N
Synonym cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
PubChem CID 2906
IUPAC Name cyclopentanamine
SMILES NC1CCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0196614
View Documents
Thermo Scientific Chemicals
L0196614
25 g
Each for $76.63
 
AAL0196622
View Documents
Thermo Scientific Chemicals
L0196622
100 g
Each for $204.41
 
Description

Description

Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

Solubility
Miscible with water, acetone and benzene.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and carbon dioxide.
Specifications

Chemical Identifiers

1003-03-8
85.15
NISGSNTVMOOSJQ-UHFFFAOYSA-N
2906
NC1CCCC1
C5H11N
MFCD00001380
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
cyclopentanamine

Specifications

1003-03-8
0.862
106°C to 108°C
Amine-like
1.4505
25 g
635706
Miscible with water,acetone and benzene.
NC1CCCC1
85.15
85.15
Cyclopentylamine
-85°C
12.5
12°C (53°F)
C5H11N
MFCD00001380
UN3286
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
NISGSNTVMOOSJQ-UHFFFAOYSA-N
cyclopentanamine
2906
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H331-H314-H318-H317
Highly flammable liquid and vapor.
Fatal if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H225-H301-H314-H317-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.

EINECSNumber : 213-697-3

RTECSNumber : GY8452000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only