missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Cyclopropanecarboxamidoxime, 97%
Supplier: Alfa Aesar™ H5823604
Specifications
Cyclopropanecarboxamidoxime | |
C4H8N2O | |
2g | |
OMCUPXRCMTUDHI-UHFFFAOYSA-N | |
N'-hydroxycyclopropanecarboximidamide | |
9582826 | |
97% |
51285-13-3 | |
MFCD03426266 | |
n-hydroxycyclopropanecarboximidamide, z-n'-hydroxycyclopropanecarboxamidine, n-hydroxycyclopropanecarboxamidine, n'-hydroxycyclopropanecarboxamidine, z-n'-hydroxycyclopropanec arboximidamide, cyclopropanecarboxamidoxime, n-hydroxy-cyclopropanecarboximidamide, n'-hydroxycyclopropane-carboximidamide, z-n'-hydroxycyclopropanecarboximidamide, n'-hydroxycyclopropanecarboximideamide | |
C1CC1C(=NO)N | |
100.121 | |
100.12 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning