Learn More
Cyclopropanemethylamine, 97%, Thermo Scientific Chemicals
$96.04 - $966.05
Chemical Identifiers
CAS | 2516-47-4 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.123 |
MDL Number | MFCD00037147 |
InChI Key | IGSKHXTUVXSOMB-UHFFFAOYSA-N |
Synonym | cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine |
PubChem CID | 75646 |
IUPAC Name | cyclopropylmethanamine |
SMILES | C1CC1CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1394603
|
Thermo Scientific Chemicals
A1394603 |
1 g |
Each for $96.04
|
|
||||||
AAA1394606
|
Thermo Scientific Chemicals
A1394606 |
5 g |
Each for $257.49
|
|
||||||
AAA1394614
|
Thermo Scientific Chemicals
A1394614 |
25 g |
Each for $966.05
|
|
||||||
Description
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
Chemical Identifiers
2516-47-4 | |
71.123 | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
75646 | |
C1CC1CN |
C4H9N | |
MFCD00037147 | |
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
cyclopropylmethanamine |
Specifications
2516-47-4 | |
85°C to 86°C | |
Amine-like | |
1.433 | |
1 g | |
773674 | |
cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
cyclopropylmethanamine | |
75646 | |
≥98% |
0.82 | |
2°C (36°F) | |
C4H9N | |
MFCD00037147 | |
UN2733 | |
Air Sensitive | |
Fully miscible in water. | |
C1CC1CN | |
71.123 | |
71.12 | |
Cyclopropanemethylamine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 219-737-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only