Learn More
Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals
$86.00 - $294.01
Chemical Identifiers
CAS | 139631-62-2 |
---|---|
Molecular Formula | C3H5ClO2S |
Molecular Weight (g/mol) | 140.58 |
MDL Number | MFCD01631933 |
InChI Key | PFWWSGFPICCWGU-UHFFFAOYSA-N |
Synonym | cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride |
PubChem CID | 2760919 |
IUPAC Name | cyclopropanesulfonyl chloride |
SMILES | ClS(=O)(=O)C1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2910703
|
Thermo Scientific Chemicals
H2910703 |
1 g |
Each for $86.00
|
|
|||||
AAH2910706
|
Thermo Scientific Chemicals
H2910706 |
5 g |
Each for $294.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
139631-62-2 | |
140.58 | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
2760919 | |
ClS(=O)(=O)C1CC1 |
C3H5ClO2S | |
MFCD01631933 | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
cyclopropanesulfonyl chloride |
Specifications
139631-62-2 | |
60°C (2 mmHg) | |
C3H5ClO2S | |
MFCD01631933 | |
UN3265 | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
ClS(=O)(=O)C1CC1 | |
140.58 | |
140.59 | |
Cyclopropanesulfonyl chloride |
1.39 | |
96°C (205°F) | |
1.477 | |
1 g | |
Moisture sensitive | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
cyclopropanesulfonyl chloride | |
2760919 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only