missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl 2-Fluorobenzyl Ketone 98.0+%, TCI America™
$82.02 - $411.95
Chemical Identifiers
| CAS | 150322-73-9 |
|---|---|
| Molecular Formula | C11H11FO |
| Molecular Weight (g/mol) | 178.206 |
| MDL Number | MFCD10687164 |
| InChI Key | DWBGTJUQWKWYGB-UHFFFAOYSA-N |
| PubChem CID | 15289941 |
| IUPAC Name | 1-cyclopropyl-2-(2-fluorophenyl)ethanone |
| SMILES | C1CC1C(=O)CC2=CC=CC=C2F |
Chemical Identifiers
| 150322-73-9 | |
| 178.206 | |
| DWBGTJUQWKWYGB-UHFFFAOYSA-N | |
| 1-cyclopropyl-2-(2-fluorophenyl)ethanone |
| C11H11FO | |
| MFCD10687164 | |
| 15289941 | |
| C1CC1C(=O)CC2=CC=CC=C2F |
Specifications
| 150322-73-9 | |
| 61°C | |
| MFCD10687164 | |
| DWBGTJUQWKWYGB-UHFFFAOYSA-N | |
| 1-cyclopropyl-2-(2-fluorophenyl)ethanone | |
| 15289941 | |
| ≥98.0% (GC) | |
| Cyclopropyl 2-Fluorobenzyl Ketone |
| Yellow | |
| C11H11FO | |
| 5 g | |
| C1CC1C(=O)CC2=CC=CC=C2F | |
| 178.206 | |
| 178.21 | |
| Liquid |
Safety and Handling
TSCA : No