Learn More
Cyclopropyl phenyl ketone, 97%, Thermo Scientific Chemicals
$134.27 - $408.82
Chemical Identifiers
CAS | 3481-02-5 |
---|---|
Molecular Formula | C10H10O |
Molecular Weight (g/mol) | 146.19 |
MDL Number | MFCD00001294 |
InChI Key | PJRHFTYXYCVOSJ-UHFFFAOYSA-N |
Synonym | cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone |
PubChem CID | 77029 |
IUPAC Name | cyclopropyl(phenyl)methanone |
SMILES | C1CC1C(=O)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0221814
|
Thermo Scientific Chemicals
L0221814 |
25 g |
Each for $134.27
|
|
|||||
AAL0221822
|
Thermo Scientific Chemicals
L0221822 |
100 g |
Each for $408.82
|
|
|||||
Description
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
3481-02-5 | |
146.19 | |
PJRHFTYXYCVOSJ-UHFFFAOYSA-N | |
77029 | |
C1CC1C(=O)C2=CC=CC=C2 |
C10H10O | |
MFCD00001294 | |
cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone | |
cyclopropyl(phenyl)methanone |
Specifications
3481-02-5 | |
248°C | |
C10H10O | |
MFCD00001294 | |
1860145 | |
Insoluble in water. | |
C1CC1C(=O)C2=CC=CC=C2 | |
146.19 | |
146.19 | |
Cyclopropyl phenyl ketone |
1.06 | |
90°C (194°F) | |
1.5534 | |
25 g | |
cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone | |
PJRHFTYXYCVOSJ-UHFFFAOYSA-N | |
cyclopropyl(phenyl)methanone | |
77029 | |
97% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 222-458-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only