missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyproterone acetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 459542500

 View more versions of this product

Catalog No. AC459542500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyproterone acetate
207.0°C to 215.0°C
0.5% max. (80°C)
97.5% min. (HPLC)
250 mg
Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, methanol, slightly soluble in ethanol
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C
9880
416.94
427-51-0
0.1% max.
Authentic
C24H29ClO4
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
UWFYSQMTEOIJJG-FDTZYFLXSA-N
416.94
CHEBI:50743
98%

Chemical Identifiers

427-51-0
416.94
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
CHEBI:50743
C24H29ClO4
UWFYSQMTEOIJJG-FDTZYFLXSA-N
9880
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Warning

EINECSNumber : 207-048-3

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only