missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cystamine Dihydrochloride 97.0+%, TCI America™

Supplier:  TCI America C0875500G

 View more versions of this product

Catalog No. C0875500G


Explore available promotions

Description

Description

Specifications

Specifications

Cystamine Dihydrochloride
218°C
C4H14Cl2N2S2
500 g
YUFRRMZSSPQMOS-UHFFFAOYSA-N
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
5941
≥97.0% (N)
56-17-7
White
MFCD00012905
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
C(CSSCCN)N.Cl.Cl
225.19
225.19
Crystalline Powder

Chemical Identifiers

56-17-7
225.19
YUFRRMZSSPQMOS-UHFFFAOYSA-N
5941
C(CSSCCN)N.Cl.Cl
C4H14Cl2N2S2
MFCD00012905
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KR7260000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions