missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Cytidine-5'-diphosphate disodium salt, 95%

 

Supplier:  Thermo Scientific Chemicals J6423403

Catalog No. AAJ6423403


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cytidine-5'-diphosphate disodium salt
White
MFCD00084682
CDP disodium salt
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O
447.14
447.1
Powder
54394-90-0
C9H13N3Na2O11P2
1 g
ZGDYAAWYYNVXEB-WFIJOQBCSA-L
disodium;(2R,3R,4S,5S)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
131632379
95%

Chemical Identifiers

54394-90-0
447.14
ZGDYAAWYYNVXEB-WFIJOQBCSA-L
131632379
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O
C9H13N3Na2O11P2
MFCD00084682
CDP disodium salt
disodium;(2R,3R,4S,5S)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only